Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Butylene oxide, THF, Diethylene oxide
Stabilization:
250 ppm Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
178810-10L
178810-1L
178810-2.5L
178810-20L
BJ178810-10LEA
507.46
USD
BJ178810-10L
BJ178810-1L
BJ178810-2.5L
BJ178810-20L
Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade
Tetrahydrofuran
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 66 Melting Pt: -108.5 Density: 0.89 g/cm³ (20 °C) Flash Pt: –21 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005356 CAS Number: 109-99-9 EINECS: 203-726-8 UN: 2056 ADR: 3 II Merck Index: 12,09351 |
|
Specification Test Results
Identity (IR) | complying |
Assay (GC) | Min. 99.0 % |
Assay BHT (GC) | 200 - 300 ppm |
APHA | Max. 20 |
Refractive Index | n20/D 1.407 (lit.) |
Explosion Limit | 11.8 % |
Vapor Pressure | 114 mmHg ( 15 °C) |
Vapor Density | 2.5 (vs air) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...