Bee Apamin

Supplier: AnaSpec
AS-60772
103005-972EA 214.85 USD
103005-972
Bee Apamin
Proteins and Peptides
A 18-amino acid peptide from bee venom, a selective blocker of calcium activated potassium channels.
Sequence:CNCKAPETALCARRCQQH-NH2 (Disulfide bridge: 1-11; 3-15)
MW:2027.4 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR