IPTG (isopropyl-β-D-thiogalactopyranoside) 0.5 M in water ≥98%

Supplier: Zymo Research
Warning

Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside

I1001-25 I1001-5
77001-958EA 50.93 USD
77001-958 77001-960
IPTG (isopropyl-β-D-thiogalactopyranoside) 0.5 M in water ≥98%
IPTG (isopropyl-β-D-thiogalactopyranoside)

Premade IPTG in water.


  • Purity: 98%
  • Concentration: 0.5 M
  • Storage: –20 °C

Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR