Human Fibrinolysis Inhibiting Factor

Supplier: AnaSpec
AS-24262
102999-798EA 185.62 USD
102999-798
Human Fibrinolysis Inhibiting Factor
Proteins and Peptides
This peptide mimics the N terminus of the alpha-chain of fibrin. It binds to the 'a' polymerization pocket in the alpha-chain more tightly than a peptide corresponding to the chain native sequence GPRV. The complex between GPRP and fibrinogen causes an increased resistance to degradation by plasmin, and the polymerization reaction is depressed by the addition of excess peptide. Fibrin assembly, as well as the acceleration of the factor XIIIa reaction, can be prevented by this homologue of the natural sequence of amino acids.
Sequence:GPRP
MW:425.5 Da
%Peak area by HPLC:≥95%
Storage condition:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR