LiChrosorb® Sorbents, MilliporeSigma, Merck
Supplier: MilliporeSigma
1.09309.0010
1.09334.0010
1.09318.0010
1.09332.0010
1.09333.0010
10143-782EA
407.86
USD
10143-782
10143-784
10144-012
10144-014
10144-016
LiChrosorb® Sorbents, MilliporeSigma, Merck
Chromatography Sorbents
HPLC Sorbents
LiChrosorb® Si 100 is a reliable irregular shaped silica sorbent with a pore diameter of 100 Å for normal phase chromatography.
LiChrosorb® RP-8 is a reliable irregular shaped silica gel carrier with reversed phase properties. It is well suited for the separation of neutral, acidic and weak basic compounds.
LiChrosorb® RP-18 is a reliable irregular shaped silica gel carrier with reversed phase properties. It is well suited for the separation of neutral, acidic and weak basic compounds.