I125 Pico-Calibrator, Revvity
5080225 5080125 5080525 5081025
101679-676PK 3592.31 USD
101679-676 101677-082 101683-042 101689-874
I125 Pico-Calibrator, Revvity
Scintillation Cocktails
Pico-Calibrators are matched sets of radionuclide (gamma) point sources that are sealed with epoxy in 12 x75 mm polypropylene tubes.
  • Isotope-I-125
Activity is verified using a NaI detector for which the efficiency has been established using NIST traceable standards. Each source is labeled to identify the radionuclide, activity (in both microCuries and DPM), assay date and lot number. Standards are made in groups which are matched by counts to ensure +/- 1% source matching. Pico-Calibrators are typically used for verifying detector counting efficiency and normalizing multi-detector gamma counters. The calibrators are supplied with a Pico-Calibrator log book that contains technical information as well as a template for quality assurance.
Información para pedidos: This is a radioactive product - shipping address must have a license to receive radioactive materials.