Hydrogen Peroxide Cell-Based Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical Company
600050-480 600050-96
75817-302EA 444.93 USD
75817-302 75817-304
Hydrogen Peroxide Cell-Based Assay Kit, Cayman Chemical Company
Assays Cellular Assays
A fluorometric detection assay for hydrogen peroxide.
  • A simple fluorometric assay for the quantification of extracellular H2O2
  • Utilizes ADHP, a sensitive and stable probe for H2O2
  • Catalase, an H2O2 scavenger, is included as a control for specificity
  • Rapid assay; results in under 1 hour
  • Midpoint: 50% B/B0: 100 – 300 pg/mL
Reactive oxygen species, such as hydrogen peroxide (H2O2) and superoxide are generated by phagocytes and participate in damaging invading miccroorganisms or other biologic targets. Cayman's H2O2 Cell-Based Assay Kit provides a simple fluorometric method for the sensitive quantitation of extracellular H2O2 produced by cultured cells. H2O2 is detected using ADHP (10-acetyl-3,7-dihydroxyphenoxazine), a highly sensitive and stable probe for H2O2. In the presence of horseradish peroxidase, ADHP reacts with H2O2 with a 1:1 stoichiometry to produce highly fluorescent resorufin. Resorufin fluorescence can be read using an excitation wavelength between 530 – 560 nm and an emission wavelength of 590 nm. Catalase, an H2O2 scavenger, is included in the kit for checking specificity of the assay.