1,4,7,10-Tetraazacyclododecane ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Cyclen

07-1941-25G 07-1941-1G 07-1941-5G
100203-710EA 634.09 USD
100203-710 100197-330 100207-334
1,4,7,10-Tetraazacyclododecane ≥98%
1,4,7,10-Tetraazacyclododecane
Formula: C₈H₂₀N₄
MW: 172.27 g/mol
MDL Number: MFCD00066281
CAS Number: 294-90-6

Order Now

Specification Test Results

Color and Form White xtl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR