Flame Resistant Coverall, Khaki, Westex DH, Lakeland Industries
Supplier: Lakeland
C065DH20-36-30 C065DH20-40-30 C065DH20-44-30 C065DH20-48-30 C065DH20-52-30 C065DH20-56-30 C065DH20-60-30 C065DH20-64-30
75923-446EA 267.89 USD
75923-446 75923-448 75923-450 75923-452 75923-454 75923-456 75923-458 75923-460
Flame Resistant Coverall, Khaki, Westex DH, Lakeland Industries
Workwear
Inherently FR Coverall for primary protection from flash fire and electrical arc flash.
  • Inherently FR fabric that can't lose its flame resistant properties
  • NFPA 2112, UL Certified for Flash Fire Protection
  • NFPA 70E, CAT2 Electrical Arc Flash Protection
  • Pleated back and elastic waist for fit and comfort
  • Flared leg cuffs for ease of donning and doffing
Westex DH Flame Resistant Coverall, dual certified for both flash fire and electrical arc flash protection. Designed for fit and comfort with pleated back, elastic waist, and plenty of pockets. Flared leg openings make donning and doffing easier, and sleeve cuffs feature adjustable snap closures.
Certificaciones: NFPA 2112. NFPA 70E, CAT2.