BD BBL™ Fluid Thioglycollate Medium Bottled Media, BD Diagnostics
Supplier: BD
257144
292788
BD257144PK
627.12
USD
BD257144
95008-288
BD BBL™ Fluid Thioglycollate Medium Bottled Media, BD Diagnostics
Media for Microbiology
Prepared Media for Microbiology
BD fluid thioglycollate medium is used for the sterility testing of biologics and for the enrichment and cultivation of anaerobes, aerobes, and microaerophiles.