Accessories for Super Erecta® Shelving Systems, Metro International
Supplier: Metro
R61S SAKITA2 R84S R61C 9084Z
19766-890EA 67.15 USD
19766-890 97057-282 19766-894 19766-898 19766-904
Accessories for Super Erecta® Shelving Systems, Metro International
Furniture Shelves
Form side and back enclosures for a shelving unit
Can also serve as uniform dividers within unit by passing through shelves from top to bottom. Recommended spacing is every 6" (150 mm) to 12" (305 mm), depending upon shelf contents.
Información de suministro: Rods are shipped with four tabs per rod.