BZK Antiseptic Wipes, First Aid Only
Supplier: First Aid Only
FAE-4002-001 H307-002 AN337-10
76049-096BX 9.79 USD
76049-096 76049-238 76049-246
BZK Antiseptic Wipes, First Aid Only
Wipes Disposable Wipes and Towels Disinfectant Wipes
First Aid antiseptic wipes to help prevent infection in minor cuts, scrapes, minor burns, and for cleaning skin without the use of soap and water.
  • Cleanse and prevent infection in minor wounds, burns and scrapes
  • Saturated with 0.133% Benzalkonium Chloride
  • Individual packets are easy and convenient to use
  • Single use packes eliminates potential cross contamination
  • Substitute for cleaning skin without the use of soap and water
  • Perfect as a first aid refill or standalone supply
Each pad is saturated with 0.133% Benzalkonium Chloride to help prevent infection in minor cuts, burns and scrapes. The wipes can also be used as a substitute for cleaning skin without the use of soap and water.
Individual single use packets are easy and convenient to use and eliminates potential cross contamination.
SmartCompliance refills are dispensed from the Smart Tab, ezRefill box, which includes an indicator tab to alert users when to reorder. The box snaps easily into place in all SmartCompliance cabinets, keeping supplies organized. For use with the following SmartCompliance systems: 90608, 90580, 90659, 90660, 746000, 746004, 746005, 746006, 90578, 90658, 90732, 90829, 90830, 90831, 90832, 90833, 90834 90835, 1000-FAE-0103, 1001-FAE-0103, 1300-FAE-0103, 1301-FAE-0103, 1050-FAE-0103, 1350-FAE-0103.