L-α-phosphatidylinositol-4,5-bisphosphate (from porcine) ≥99% (by TLC) Ammonium salt

Supplier: AVANTI POLAR LIPIDS INC MS

Synonyms: Brain PI(4,5)P2

840046X25MG 840046X1MG 840046X10MG 840046X5MG
100126-690EA 12582.46 USD
100126-690 100128-770 100121-068 100123-350
L-α-phosphatidylinositol-4,5-bisphosphate (from porcine) ≥99% (by TLC) Ammonium salt
L-α-phosphatidylinositol-4,5-bisphosphate

PI(4,5)P2 plays an important role as a substrate in various signalling proteins though it is a minor component in cell membranes


  • Liquid


As a subsrate in PI 3-kinase, PI(4,5)P2 is phosphorylated to PI(3,4,5)P3 which act as an activator for downstream signaling components such as the protein kinase AKT.


This is an intermediate in the IP3/DAG pathway where it is hydrolyzed by phospholipase C to liberate the second messengers, inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG).

MW: 1096.385 g/mol
Storage Temperature: Freezer
CAS Number: 383907-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR