Peroxide-Cured Versilic™ Silicone Tubing, Chemglass
Supplier: Chemglass
CLS-1436-T20 CLS-1436-T21 CLS-1436-T22 CLS-1436-T23 CLS-1436-T24 CLS-1436-T25
30622-854EA 81.48 USD
30622-854 30622-856 30622-858 30622-860 30622-862 30622-864
Peroxide-Cured Versilic™ Silicone Tubing, Chemglass
Tubing Transfer Tubing
Peroxide-cured Versilic™ silicone tubing is designed for use in applications where flexibility, resiliency and durability are required.
  • Non-toxic, non-hemolytic and non-pyrogenic
  • Temperature extremes from –75 to 350 °F
  • Standard lengths of 50'
Certificaciones: Tubing complies fully with the requirements of the USP Class VI Criteria. It also meets 3-A Sanitary Standards, FDA 21 CFR Part 177.2600 Criteria and NSF 51 Standard.