Picric acid, (∼30% H₂O) ≥99% (by GC) ACS

Supplier: CHEM IMPEX INTL INC MS
Danger

Synonyms: 2,4,6-Trinitrophenol, TNP

00312-500G 00312-500G
102572-740EA 292.31 USD
102572-740 CH00312-500G
Picric acid, (∼30% H₂O) ≥99% (by GC) ACS
Picric acid
Formula: C₆H₃N₃O₇
Melting Pt: 121…123 °C
Density: 1.8
Flash Pt: 150 °C
MDL Number: MFCD00007102
CAS Number: 88-89-1
UN: 1344
ADR: 4.1,I

Order Now

Specification Test Results

Molecular Formula C6H3N3O7
Appearance Moist yellow powder
Melting Point 121 - 123 °C
Flash Point 150 °C
Density 1.8
Sulfate (SO42−) ≤0.1%
Water Content (Karl Fisher) ≥30%
pKa 0.4
Insoluble ≤ 0.1% (in Toluene)
Insoluble material ≤ 0.01% (Resinous matter)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR