QUIPTROL 3000 Chlorine-Dioxide Cleaner, Quip Laboratories
Supplier: Quip Laboratories
Q3000-15GAL Q3000-30GAL Q3000-55GAL Q3000-5GAL
10121-254EA 2173.04 USD
10121-254 10121-256 10121-258 10121-260
QUIPTROL 3000 Chlorine-Dioxide Cleaner, Quip Laboratories
Cleaning Agents Disinfectants
Quiptrol 3000 is a liquid chlorine dioxide concentrate that can be applied in one step, at room temperature, to water via simple chemical feed pump.
  • Ready to use
It is perfect for sanitizing process water, piping systems, cooling towers, condensers and heat exchangers because it removes biofilm by penetrating and destroying microorganisms safely, quickly and simply.
Certificaciones: Approved by U.S EPA for water purification. Certified by the National Sanitary Foundation under the NSF/ANSI Standard 60 for drinking water. Registered with the U.S. EPA-FIRFA as an anti-microbial pesticide (EPA Reg. No. 75757-2)