Polyethylene glycol 3,350 50% (w/v)

Supplier: Rigaku Reagents

Synonyms: PEG 3,350, PEG 3350

1008054 1008055
101443-470EA 103.31 USD
101443-470 101443-862
Polyethylene glycol 3,350 50% (w/v)
Polyethylene glycol 3,350

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: H(OCH₂CH₂)nOH
MW: 3350 g/mol
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR