Sodium chloride 5 M

Supplier: Rigaku Reagents

1008390 1008391
101443-388EA 79.77 USD
101443-388 101443-748
Sodium chloride 5 M
Sodium chloride

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: NaCl
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR