Potassium thiocyanate 1 M

Supplier: Rigaku Reagents

1008215 1008214
101443-688EA 162.15 USD
101443-688 101443-340
Potassium thiocyanate 1 M
Potassium thiocyanate

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: KSCN
MDL Number: MFCD00011413
CAS Number: 333-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR