Potassium sulfate 0.5 M
Supplier: Rigaku Reagents
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.
Formula:
K₂SO₄ |
MDL Number:
MFCD00011388 CAS Number: 7778-80-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...