di-Potassium hydrogen phosphate 3 M

Supplier: Rigaku Reagents

1008361 1008360
101443-662EA 149.08 USD
101443-662 101443-314
di-Potassium hydrogen phosphate 3 M
di-Potassium hydrogen phosphate

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: K₂HPO₄
MDL Number: MFCD00011383
CAS Number: 7758-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=2fa248735fc54d9c7253a066a62570d2de7b7ff3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR