Tripotassium citrate 1 M

Supplier: Rigaku Reagents

1008213 1008212
101443-516EA 129.46 USD
101443-516 101443-204
Tripotassium citrate 1 M
Tripotassium citrate solutions

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: HOC(COOK)(CH₂COOK)₂·H₂O
MW: 324.41 g/mol
MDL Number: MFCD00150442
CAS Number: 6100-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR