Ammonium dihydrogen phosphate 1 M

Supplier: Rigaku Reagents

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

1008106 1008105
101443-826EA 112.46 USD
101443-826 101443-438
Ammonium dihydrogen phosphate 1 M
Ammonium dihydrogen phosphate

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 190
MDL Number: MFCD00003396
CAS Number: 7722-76-1
Merck Index: 13,00547

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR