KWIK-STIK 2 Pack, Microbiologics
Supplier: MICROBIOLOGICS, INC.
01108P 01110P 01112P 01111P 01122P 01113P 5244P 5245P 5246P 5247P 5248P 5249P 5250P 01134P 01130P 01140P 01175P 01188P 01173P 01170P 01209P 01189P 01150P 01214P 01182P 01167P 01132P 01139P 01185P 01186P 01169P 01196P 01172P 01184P 01202P 01143P 01204P 01229P 01193P 01192P 01136P 01171P 01117P 01148P 01190P 01181P 01142P 01149P 01200P 01133P 01187P 01152P 01159P 01197P 01141P 01233P 01234P 01235P 01236P 01237P 01238P 01256P 01244P 01245P 01247P 01257P 01260P 01263P 01259P 01261P 01262P 01265P 01266P
10119-764EA 110.52 USD
10119-764 10119-780 10120-218 10127-612 10127-694 10127-704 75878-388 75878-390 75878-392 75878-394 75878-396 75878-398 75878-400 75878-668 75878-672 75878-676 75878-682 75878-802 75878-806 75878-818 75878-822 75878-826 75878-830 75878-834 75878-842 75878-850 75878-858 75878-862 75878-866 75878-874 75878-882 75878-886 75878-890 75878-898 75878-906 75878-910 75878-918 75878-922 75878-926 75878-930 75878-946 75878-950 75878-954 75878-958 75878-968 75878-972 75878-980 75878-984 75878-992 75879-000 75879-004 75879-008 75879-012 75879-044 75879-048 75879-056 75879-064 75879-076 75879-080 75879-092 76199-770 76199-774 76199-778 76199-782 76199-786 76199-790 76199-796 76199-822 76199-824 76199-826 76199-834 76199-844 76199-924 76291-822 76291-828 76291-884 76291-886 76291-890 76299-444 76299-458
KWIK-STIK 2 Pack, Microbiologics
Controls and Calibration Verification Materials
Everything you need to grow reference cultures for QC testing is included in this one handy device.
  • Over 900 strains available
  • All-in-one design reduces the risk of contamination
  • Strains are less than or equal to four passages from reference culture
  • Ready-to-use format saves time and money
  • Refrigerated storage is easy and economical
  • Online certificate of analysis provides detailed strain information
  • Traceability to reference cultures ensures authenticity
  • Technical support experts available for guidance
KWIK-STIK’s simple, all-inclusive design makes life easier for lab techs while reducing chances for errors. Each KWIK-STIK contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab. Everything you need to grow reference cultures for QC testing is included in this one handy device. Available in a two pack, universally used for all types of microbiological control testing.
Información para pedidos: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the website. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.