Epower™, Microbiologics
Supplier: MICROBIOLOGICS, INC.
0488E7 0699E3 01192E3 0443E4
75878-426EA 515.81 USD
75878-426 75879-210 76218-710 76218-712
Epower™, Microbiologics
Controls and Calibration Verification Materials
You’ll love the versatility of simply using one product for all of your QC needs – from media QC and presence/absence testing to enumeration testing, water testing, and disinfectant testing.
Available in concentrations ranging from 102 and 108 CFU per pellet.
Strains are less than or equal to four passages from reference culture.
Can be combined for a mixed microorganism population.
Easily manipulated to deliver a wide variety of concentrations.
Ready-to-use format to save you time and money.
Refrigerated storage is easy and economical.
Traceability to reference cultures ensures authenticity.
Online Certificate of Analysis provides detailed strain information.
Technical Support experts available for guidance.
Epower is the embodiment of “flexibility is your friend”. You’ll love the versatility of simply using one product for all of your QC needs – from media QC and presence/absence testing to enumeration testing, water testing, and disinfectant testing. Use a single strain or combine multiple strains for a mixed population. Each quantitative microorganism pellet delivers a specific range of CFU, so you can easily manipulate Epower to deliver virtually any concentration. We know you’ll appreciate the ease and adaptability that Epower offers.
Información para pedidos: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the website. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.