Gamma Wipe® 120 Cleanroom Laundered Wipes, Berkshire
Supplier: Berkshire
GW120ST15 GW120ST0940
21913-811CS 848.79 USD
21913-811 76097-928
Gamma Wipe® 120 Cleanroom Laundered Wipes, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Each package of Gamma Wipe® 120 wipes is gamma irradiated sterile to a 10⁻⁶ sterility assurance level. This combination of properties provides them with the high level of cleanliness, abrasion resistance, and chemical compatibility required for cleaning critical sterile processing environments.
  • Low particles, fibers, ions, and extractables
  • High absorbency
  • High abrasion resistance
  • Soft hand and edge for sensitive surfaces
  • Chemically compatible with common cleaning and disinfecting solutions, including isopropyl alcohol, phenols, bleaches, and quarternary ammonium compounds
Wipes are a knife-cut, high absorbency cleanroom laundered wiper recommended for ISO Class 4 and above environments. The wipes are composed of 100% continuous filament polyester knit fabric.