LB Agar Plates, Teknova
Supplier: TEKNOVA INC
L1100 L7200
100216-614PK 120.11 USD
100216-614 100217-280 100220-120 100218-472 100218-252 77443-316 77528-992
LB Agar Plates, Teknova
Media for Microbiology Prepared Media for Microbiology
For the growth of E. coli and other enteric bacteria.
  • Sleeve of 20 plates
  • Sterile
Ingredients: 1.0% Tryptone (10.0 g/L), 0.5% Yeast Extract (5.0 g/L), 1.0% Sodium chloride (NaCl) (10.0 g/L), 1.5% Agar (15.0 g/L).
Ambient shipping conditions. Store at room temperature.