Betaine aldehyde chloride ≥99% (by TLC), Sigma-Aldrich®

B3650-10MG B3650-2MG B3650-100MG
220111-575EA 230.22 USD
220111-575 220111-576 220111-574
Betaine aldehyde chloride ≥99% (by TLC), Sigma-Aldrich®
Betaine aldehyde chloride

Synonym: (Formylmethyl)trimethylammonium chloride.

Formula: C₅H₁₂ClNO
MDL Number: MFCD00063509
CAS Number: 7758-31-8

Order Now

Specification Test Results

Appearance (color) White to Off-White
Appearance (Form) Powder
Solubility (Color) Colorless
Solubility (Turbidity) Clear
Water (by Karl Fischer) ≤15.0%
Carbon (anhydrous) 42.0 - 44.0%
Nitrogen (anhydrous) 9.6 - 10.3%
Infrared spectrum Conforms to Structure
Purity (TLC) ≥99%
Storage Temperature 2 - 8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR