Human Recombinant GALECTIN-1 (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-30AF-10UG 100-30AF-50UG 100-30AF-100UG 100-30AF-500UG 100-30AF-1MG
76046-738EA 90.25 USD
76046-738 76046-740 76046-742 76046-744 76046-746
Human Recombinant GALECTIN-1 (Animal free) (from E. coli)
Proteins and Peptides
Neurotrophin-4 (NT-4) is an important member of the nerve growth factor (NGF) family of proteins. Neurotrophins undergo paracrine and autocrine signaling to control neuronal survival, neuronal differentiation, and dendrite outgrowth. NT-4 is expressed ubiquitously and signals through the TrkB receptor tryrosine kinase.
  • High quality
  • Low endotoxin
  • Affordable
Galectin-1 belongs to the lectin family of carbohydrate binding proteins and binds glycans as both a monomer and a homodimer. Galectin-1 is produced in peripheral lymphoid organs and inflammatory sites. Galectin-1 plays important roles in acute and chronic inflammatory processes, cell growth, cell proliferation, and induces apoptosis of activated T cells. Galectin-1 also modulates cytokine secretion and inhibits pro-inflammatory cytokine production. 
Precaución: For research use only.