Human Recombinant FGF-Acidic (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-29AF-10UG
100-29AF-50UG
100-29AF-100UG
100-29AF-500UG
100-29AF-1MG
76046-728EA
83.09
USD
76046-728
76046-730
76046-732
76046-734
76046-736
Human Recombinant FGF-Acidic (from E. coli)
Proteins and Peptides
Acidic fibroblast growth factor (FGF-acidic), also known as FGF-1, is a potent inducer of DNA synthesis, cell proliferation, and has chemotactic activities. FGF-acidic regulates cardiogenesis through protein kinase C signaling. FGF-acidic also functions as an insulin sensitizer and mediates adipose tissue remodeling. High serum levels of FGF-acidic are associated with type 2 diabetes mellitus (T2DM), suggesting a pathogenic role of FGF-acidic during T2DM.
- High quality
- Low endotoxin
- Affordable