Stearic acid, [9,10-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT988-1MCI MT988-5MCI
101974-066EA 3164.62 USD
101974-066 101974-068
Stearic acid, [9,10-3H] ≥97% (by HPLC), MORpure™
Stearic acid, [9,10-3H]
CAS Number: 13057-51-7
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR