Penciclovir, [8-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT972-250UCI MT972-1MCI
101974-048EA 2700.38 USD
101974-048 101974-052
Penciclovir, [8-3H] ≥97%, MORpure™
Penciclovir, [8-3H]
CAS Number: 285987-74-8
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR