7-(S)-Brefeldin A, [7-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT966-250UCI MT966-1MCI
101974-032EA 2700.38 USD
101974-032 101974-034
7-(S)-Brefeldin A, [7-3H] ≥97%, MORpure™
7-(S)-Brefeldin A, [7-3H]
CAS Number: 313989-87-6
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR