Water, [3H] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT924E-1CI MT924E-5CI MT924E-10CI
101973-992EA 1798.08 USD
101973-992 101973-994 101973-996
Water, [3H] ≥98%, MORpure™
Water, [3H]
CAS Number: 13670-17-2
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR