Tetracycline, [7-3H] ≥95%

Supplier: MORAVEK BIOCHEMICALS MS

MT907-250UCI MT907-1MCI MT907-5MCI
101973-898EA 2700.38 USD
101973-898 101973-900 101973-902
Tetracycline, [7-3H] ≥95%
Tetracycline, [7-3H]
CAS Number: 2087439-71-0
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR