Acetyl coenzyme A, [acetyl-3H] ≥95% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT896H-50UCI MT896H-250UCI MT896H-1MCI
101973-854EA 1464.62 USD
101973-854 101973-856 101973-858
Acetyl coenzyme A, [acetyl-3H] ≥95% (by HPLC)
Acetyl coenzyme A, [acetyl-3H]
CAS Number: 2087954-20-7
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR