Acetyl coenzyme A, [3H] ≥95% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT896-50UCI MT896-250UCI
101973-848EA 1464.62 USD
101973-848 101973-850
Acetyl coenzyme A, [3H] ≥95% (by HPLC)
Acetyl coenzyme A, [3H]
CAS Number: 83677-16-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR