L-Tyrosine, [ring-3,5-3H] ≥96%

Supplier: MORAVEK BIOCHEMICALS MS

MT887-250UCI MT887-1MCI MT887-5MCI
101973-842EA 2700.38 USD
101973-842 101973-844 101973-846
L-Tyrosine, [ring-3,5-3H] ≥96%
L-Tyrosine, [ring-3,5-3H]
CAS Number: 7234-03-9
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR