L-Glutamic acid, [3,4-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT855-250UCI MT855-1MCI MT855-5MCI
101973-796EA 2700.38 USD
101973-796 101973-798 101973-800
L-Glutamic acid, [3,4-3H] ≥97%, MORpure™
L-Glutamic acid, [3,4-3H]
CAS Number: 147732-35-2
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR