2-Methylthioadenosine-5'-diphosphate, triammonium salt, [8-3H] ≥96%

Supplier: MORAVEK BIOCHEMICALS MS

MT850-250UCI MT850-1MCI
101973-782EA 2700.38 USD
101973-782 101973-784
2-Methylthioadenosine-5'-diphosphate, triammonium salt, [8-3H] ≥96%
2-Methylthioadenosine-5'-diphosphate, triammonium salt, [8-3H]
CAS Number: 340130-20-3
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR