Thymidine (2'-deoxythymidine)

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine

MT846W-250UCI MT846W-1MCI MT846W-5MCI
101973-758EA 2700.38 USD
101973-758 101973-760 101973-762
Thymidine (2'-deoxythymidine)
Thymidine (2'-deoxythymidine)
Formula: C₁₀H₁₄N₂O₅
MDL Number: MFCD00006537
CAS Number: 50-89-5
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR