Guanine, [8-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT839-50UCI MT839-250UCI MT839-1MCI
101973-730EA 1601.92 USD
101973-730 101973-732 101973-734
Guanine, [8-3H] ≥97%, MORpure™
Guanine, [8-3H]
CAS Number: 23199-21-5
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR