Cidofovir, [5-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT833-50UCI MT833-250UCI MT833-1MCI
101973-718EA 1621.54 USD
101973-718 101973-720 101973-722
Cidofovir, [5-3H] ≥97% (by HPLC), MORpure™
Cidofovir, [5-3H]
CAS Number: 339532-39-7
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR