Adenine, [2,8-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT789-250UCI MT789-1MCI MT789-5MCI
101973-630EA 2700.38 USD
101973-630 101973-632 101973-634
Adenine, [2,8-3H] ≥97% (by HPLC), MORpure™
Adenine, [2,8-3H]
CAS Number: 72093-28-8
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR