Etoposide, [3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT786-250UCI MT786-500UCI MT786-1MCI
101973-618EA 2700.38 USD
101973-618 101973-620 101973-622
Etoposide, [3H] ≥97%, MORpure™
Etoposide, [3H]
CAS Number: 339529-08-7
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR