Uridine, [5'-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT784-250UCI MT784-1MCI
101973-608EA 2700.38 USD
101973-608 101973-610
Uridine, [5'-3H] ≥97% (by HPLC), MORpure™
Uridine, [5'-3H]
CAS Number: 89457-59-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR