Thymidine-5'-triphosphate, tetraammonium salt, [methyl-3H] ≥95%

Supplier: MORAVEK BIOCHEMICALS MS

MT781A-250UCI MT781A-1MCI MT781A-5MCI
101973-574EA 2700.38 USD
101973-574 101973-576 101973-578
Thymidine-5'-triphosphate, tetraammonium salt, [methyl-3H] ≥95%
Thymidine-5'-triphosphate, tetraammonium salt, [methyl-3H]
CAS Number: 2088103-40-4
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR