Thymidine-5'-triphosphate, tetraammonium salt, [methyl-3H] ≥95%

Supplier: MORAVEK BIOCHEMICALS MS

MT781-250UCI MT781-1MCI MT781-5MCI
101973-568EA 2700.38 USD
101973-568 101973-570 101973-572
Thymidine-5'-triphosphate, tetraammonium salt, [methyl-3H] ≥95%
Thymidine-5'-triphosphate, tetraammonium salt, [methyl-3H]
CAS Number: 2088103-40-4
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR