Formycin B, [3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT771-500UCI MT771-1MCI
101973-564EA 4256.54 USD
101973-564 101973-566
Formycin B, [3H] ≥97%, MORpure™
Formycin B, [3H]
CAS Number: 316351-59-4
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR