2-Fluoroadenine-β-D-arabinofuranoside, [8-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT740-250UCI MT740-1MCI
101973-528EA 2700.38 USD
101973-528 101973-530
2-Fluoroadenine-β-D-arabinofuranoside, [8-3H] ≥97%, MORpure™
2-Fluoroadenine-β-D-arabinofuranoside, [8-3H]
CAS Number: 339527-33-2
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR